Assay Details
Gene Symbol: LOC5576879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847803.1 | XP_021703495.1 | 3 - 4 | 836 | 72 | |
XM_021847810.1 | XP_021703502.1 | 2 - 3 | 908 | 72 | ||
GenBank mRNA | AY432129.1 | - | 1 - 2 | 203 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
