Assay Details
Gene Symbol: LOC5578276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663754.2 | XP_001663804.1 | 2 - 3 | 538 | 104 | |
GenBank mRNA | AY432204.1 | - | 1 - 2 | 288 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
