Assay Details
Gene Symbol: LOC5567314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651699.2 | XP_001651749.1 | 1 - 2 | 254 | 77 | |
GenBank mRNA | AY432286.1 | - | 1 - 2 | 165 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
