Assay Details
Gene Symbol: LOC5567314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651699.2 | XP_001651749.1 | 2 - 3 | 379 | 62 | |
GenBank mRNA | AY432286.1 | - | 2 - 3 | 290 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
