Assay Details
Gene Symbol: LOC5563726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001647961.2 | XP_001648011.1 | 1 - 2 | 401 | 108 | |
GenBank mRNA | AY432292.1 | - | 1 - 2 | 393 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
