Assay Details
Gene Symbol: LOC5567031 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651374.2 | XP_001651424.1 | 5 | 1523 | 112 | |
GenBank mRNA | AY432296.1 | - | 1 | 216 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
