Assay Details
Gene Symbol: LOC5578301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656833.2 | XP_001656883.2 | 1 - 2 | 339 | 99 | |
XM_021851465.1 | XP_021707157.1 | 1 - 2 | 339 | 99 | ||
GenBank mRNA | AY432398.1 | - | 1 - 2 | 312 | 99 | |
AY552054.1 | - | 1 - 2 | 304 | 99 | ||
DQ440351.1 | - | 1 - 2 | 52 | 99 |
