Assay Details
Gene Symbol: LOC5574338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661321.2 | XP_001661371.1 | 1 - 2 | 84 | ||
GenBank mRNA | AY432456.1 | - | 1 - 2 | 210 | 84 | |
DQ440308.1 | - | 1 - 2 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
