Assay Details
Gene Symbol: LOC5575432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655700.2 | XP_001655750.2 | 3 - 4 | 332 | 70 | |
XM_001655705.2 | XP_001655755.2 | 2 - 3 | 192 | 70 | ||
XM_011494673.2 | XP_011492975.2 | 3 - 4 | 230 | 70 |

More Information
Set Membership: |
![]() ![]() |
