Assay Details
Gene Symbol: LOC5575432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655700.2 | XP_001655750.2 | 4 - 5 | 437 | 85 | |
XM_001655705.2 | XP_001655755.2 | 3 - 4 | 297 | 85 | ||
XM_011494673.2 | XP_011492975.2 | 4 - 5 | 335 | 85 |
