Assay Details
Gene Symbol: LOC5575432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655700.2 | XP_001655750.2 | 6 - 7 | 685 | 119 | |
XM_001655702.2 | XP_001655752.2 | 5 - 6 | 577 | 119 | ||
XM_001655705.2 | XP_001655755.2 | 5 - 6 | 545 | 119 | ||
XM_011494673.2 | XP_011492975.2 | 6 - 7 | 583 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
