Assay Details
Gene Symbol: LOC5568156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843214.1 | XP_021698906.1 | 1 - 2 | 259 | 83 | |
XM_021843215.1 | XP_021698907.1 | 2 - 3 | 224 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021843214.1, XM_021843215.1 transcripts.
Set Membership: |
![]() ![]() |
