Assay Details
Gene Symbol: LOC5568156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843214.1 | XP_021698906.1 | 11 - 12 | 4826 | 71 | |
XM_021843215.1 | XP_021698907.1 | 12 - 13 | 4791 | 71 |
