Assay Details
Gene Symbol: LOC5568156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843214.1 | XP_021698906.1 | 10 - 11 | 4715 | 83 | |
XM_021843215.1 | XP_021698907.1 | 11 - 12 | 4680 | 83 |
