Assay Details
Gene Symbol: LOC5568156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843214.1 | XP_021698906.1 | 6 - 7 | 3806 | 109 | |
XM_021843215.1 | XP_021698907.1 | 7 - 8 | 3771 | 109 |

More Information
Set Membership: |
![]() ![]() |
