Assay Details
Gene Symbol: LOC5569062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847326.1 | XP_021703018.1 | 10 - 11 | 6625 | 85 | |
XM_021847327.1 | XP_021703019.1 | 10 - 11 | 6004 | 85 | ||
XM_021847328.1 | XP_021703020.1 | 10 - 11 | 6648 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
