Assay Details
Gene Symbol: LOC5569062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847326.1 | XP_021703018.1 | 5 - 6 | 2216 | 100 | |
XM_021847327.1 | XP_021703019.1 | 5 - 6 | 1595 | 100 | ||
XM_021847328.1 | XP_021703020.1 | 5 - 6 | 2216 | 100 |
