Assay Details
Gene Symbol: LOC5572212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653830.2 | XP_001653880.2 | 2 - 3 | 1162 | 64 | |
XM_021843085.1 | XP_021698777.1 | 3 - 4 | 974 | 64 | ||
XM_021843086.1 | XP_021698778.1 | 3 - 4 | 928 | 64 | ||
XM_021843087.1 | XP_021698779.1 | 2 - 3 | 1162 | 64 |

More Information
Set Membership: |
![]() ![]() |
