Assay Details
Gene Symbol: LOC5572212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653830.2 | XP_001653880.2 | 5 - 6 | 4759 | 77 | |
XM_021843085.1 | XP_021698777.1 | 6 - 7 | 4571 | 77 | ||
XM_021843086.1 | XP_021698778.1 | 6 - 7 | 4525 | 77 | ||
XM_021843087.1 | XP_021698779.1 | 5 - 6 | 4759 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
