Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 4 - 5 | 1044 | 85 | |
XM_021854229.1 | XP_021709921.1 | 4 - 5 | 1031 | 85 | ||
XM_021854230.1 | XP_021709922.1 | 4 - 5 | 1019 | 85 | ||
XM_021854231.1 | XP_021709923.1 | 4 - 5 | 946 | 85 | ||
XM_021854232.1 | XP_021709924.1 | 4 - 5 | 1006 | 85 | ||
XM_021854233.1 | XP_021709925.1 | 4 - 5 | 940 | 85 | ||
XM_021854234.1 | XP_021709926.1 | 2 - 3 | 649 | 85 | ||
XM_021854235.1 | XP_021709927.1 | 4 - 5 | 942 | 85 | ||
XM_021854236.1 | XP_021709928.1 | 3 - 4 | 779 | 85 | ||
XM_021854237.1 | XP_021709929.1 | 4 - 5 | 931 | 85 | ||
XM_021854238.1 | XP_021709930.1 | 4 - 5 | 843 | 85 | ||
XM_021854239.1 | XP_021709931.1 | 4 - 5 | 1497 | 85 | ||
XM_021854240.1 | XP_021709932.1 | 4 - 5 | 982 | 85 | ||
XM_021854241.1 | XP_021709933.1 | 4 - 5 | 1487 | 85 | ||
XM_021854242.1 | XP_021709934.1 | 4 - 5 | 1430 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
