Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854239.1 | XP_021709931.1 | 3 - 4 | 1364 | 102 | |
XM_021854240.1 | XP_021709932.1 | 3 - 4 | 849 | 102 | ||
XM_021854241.1 | XP_021709933.1 | 3 - 4 | 1354 | 102 | ||
XM_021854242.1 | XP_021709934.1 | 3 - 4 | 1297 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
