Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854239.1 | XP_021709931.1 | 2 - 3 | 1041 | 89 | |
XM_021854240.1 | XP_021709932.1 | 2 - 3 | 526 | 89 | ||
XM_021854241.1 | XP_021709933.1 | 2 - 3 | 1031 | 89 | ||
XM_021854242.1 | XP_021709934.1 | 2 - 3 | 974 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021854239.1, XM_021854240.1, XM_021854241.1, XM_021854242.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
