Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 15 - 16 | 3763 | 71 | |
XM_021854229.1 | XP_021709921.1 | 15 - 16 | 3750 | 71 | ||
XM_021854230.1 | XP_021709922.1 | 15 - 16 | 3738 | 71 | ||
XM_021854231.1 | XP_021709923.1 | 15 - 16 | 3665 | 71 | ||
XM_021854232.1 | XP_021709924.1 | 15 - 16 | 3725 | 71 | ||
XM_021854233.1 | XP_021709925.1 | 15 - 16 | 3659 | 71 | ||
XM_021854234.1 | XP_021709926.1 | 13 - 14 | 3368 | 71 | ||
XM_021854235.1 | XP_021709927.1 | 15 - 16 | 3661 | 71 | ||
XM_021854236.1 | XP_021709928.1 | 14 - 15 | 3498 | 71 | ||
XM_021854237.1 | XP_021709929.1 | 15 - 16 | 3650 | 71 | ||
XM_021854238.1 | XP_021709930.1 | 15 - 16 | 3562 | 71 | ||
XM_021854239.1 | XP_021709931.1 | 15 - 16 | 4216 | 71 | ||
XM_021854240.1 | XP_021709932.1 | 15 - 16 | 3701 | 71 | ||
XM_021854241.1 | XP_021709933.1 | 15 - 16 | 4206 | 71 | ||
XM_021854242.1 | XP_021709934.1 | 15 - 16 | 4149 | 71 | ||
XM_021854243.1 | XP_021709935.1 | 14 - 15 | 3566 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
