Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 14 - 15 | 3599 | 102 | |
XM_021854229.1 | XP_021709921.1 | 14 - 15 | 3586 | 102 | ||
XM_021854230.1 | XP_021709922.1 | 14 - 15 | 3574 | 102 | ||
XM_021854231.1 | XP_021709923.1 | 14 - 15 | 3501 | 102 | ||
XM_021854232.1 | XP_021709924.1 | 14 - 15 | 3561 | 102 | ||
XM_021854233.1 | XP_021709925.1 | 14 - 15 | 3495 | 102 | ||
XM_021854234.1 | XP_021709926.1 | 12 - 13 | 3204 | 102 | ||
XM_021854235.1 | XP_021709927.1 | 14 - 15 | 3497 | 102 | ||
XM_021854236.1 | XP_021709928.1 | 13 - 14 | 3334 | 102 | ||
XM_021854237.1 | XP_021709929.1 | 14 - 15 | 3486 | 102 | ||
XM_021854238.1 | XP_021709930.1 | 14 - 15 | 3398 | 102 | ||
XM_021854239.1 | XP_021709931.1 | 14 - 15 | 4052 | 102 | ||
XM_021854240.1 | XP_021709932.1 | 14 - 15 | 3537 | 102 | ||
XM_021854241.1 | XP_021709933.1 | 14 - 15 | 4042 | 102 | ||
XM_021854242.1 | XP_021709934.1 | 14 - 15 | 3985 | 102 | ||
XM_021854243.1 | XP_021709935.1 | 13 - 14 | 3402 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
