Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 13 - 14 | 3331 | 92 | |
XM_021854229.1 | XP_021709921.1 | 13 - 14 | 3318 | 92 | ||
XM_021854230.1 | XP_021709922.1 | 13 - 14 | 3306 | 92 | ||
XM_021854231.1 | XP_021709923.1 | 13 - 14 | 3233 | 92 | ||
XM_021854232.1 | XP_021709924.1 | 13 - 14 | 3293 | 92 | ||
XM_021854233.1 | XP_021709925.1 | 13 - 14 | 3227 | 92 | ||
XM_021854234.1 | XP_021709926.1 | 11 - 12 | 2936 | 92 | ||
XM_021854235.1 | XP_021709927.1 | 13 - 14 | 3229 | 92 | ||
XM_021854236.1 | XP_021709928.1 | 12 - 13 | 3066 | 92 | ||
XM_021854237.1 | XP_021709929.1 | 13 - 14 | 3218 | 92 | ||
XM_021854238.1 | XP_021709930.1 | 13 - 14 | 3130 | 92 | ||
XM_021854239.1 | XP_021709931.1 | 13 - 14 | 3784 | 92 | ||
XM_021854240.1 | XP_021709932.1 | 13 - 14 | 3269 | 92 | ||
XM_021854241.1 | XP_021709933.1 | 13 - 14 | 3774 | 92 | ||
XM_021854242.1 | XP_021709934.1 | 13 - 14 | 3717 | 92 | ||
XM_021854243.1 | XP_021709935.1 | 12 - 13 | 3134 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
