Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 12 - 13 | 3136 | 116 | |
XM_021854229.1 | XP_021709921.1 | 12 - 13 | 3123 | 116 | ||
XM_021854230.1 | XP_021709922.1 | 12 - 13 | 3111 | 116 | ||
XM_021854231.1 | XP_021709923.1 | 12 - 13 | 3038 | 116 | ||
XM_021854232.1 | XP_021709924.1 | 12 - 13 | 3098 | 116 | ||
XM_021854233.1 | XP_021709925.1 | 12 - 13 | 3032 | 116 | ||
XM_021854234.1 | XP_021709926.1 | 10 - 11 | 2741 | 116 | ||
XM_021854235.1 | XP_021709927.1 | 12 - 13 | 3034 | 116 | ||
XM_021854236.1 | XP_021709928.1 | 11 - 12 | 2871 | 116 | ||
XM_021854237.1 | XP_021709929.1 | 12 - 13 | 3023 | 116 | ||
XM_021854238.1 | XP_021709930.1 | 12 - 13 | 2935 | 116 | ||
XM_021854239.1 | XP_021709931.1 | 12 - 13 | 3589 | 116 | ||
XM_021854240.1 | XP_021709932.1 | 12 - 13 | 3074 | 116 | ||
XM_021854241.1 | XP_021709933.1 | 12 - 13 | 3579 | 116 | ||
XM_021854242.1 | XP_021709934.1 | 12 - 13 | 3522 | 116 | ||
XM_021854243.1 | XP_021709935.1 | 11 - 12 | 2939 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
