Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 9 - 10 | 2205 | 100 | |
XM_021854229.1 | XP_021709921.1 | 9 - 10 | 2192 | 100 | ||
XM_021854230.1 | XP_021709922.1 | 9 - 10 | 2180 | 100 | ||
XM_021854231.1 | XP_021709923.1 | 9 - 10 | 2107 | 100 | ||
XM_021854232.1 | XP_021709924.1 | 9 - 10 | 2167 | 100 | ||
XM_021854233.1 | XP_021709925.1 | 9 - 10 | 2101 | 100 | ||
XM_021854234.1 | XP_021709926.1 | 7 - 8 | 1810 | 100 | ||
XM_021854235.1 | XP_021709927.1 | 9 - 10 | 2103 | 100 | ||
XM_021854236.1 | XP_021709928.1 | 8 - 9 | 1940 | 100 | ||
XM_021854237.1 | XP_021709929.1 | 9 - 10 | 2092 | 100 | ||
XM_021854238.1 | XP_021709930.1 | 9 - 10 | 2004 | 100 | ||
XM_021854239.1 | XP_021709931.1 | 9 - 10 | 2658 | 100 | ||
XM_021854240.1 | XP_021709932.1 | 9 - 10 | 2143 | 100 | ||
XM_021854241.1 | XP_021709933.1 | 9 - 10 | 2648 | 100 | ||
XM_021854242.1 | XP_021709934.1 | 9 - 10 | 2591 | 100 | ||
XM_021854243.1 | XP_021709935.1 | 8 - 9 | 2008 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
