Assay Details
Gene Symbol: LOC5564524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848242.1 | XP_021703934.1 | 2 - 3 | 704 | 75 | |
XM_021848243.1 | XP_021703935.1 | 2 - 3 | 839 | 75 | ||
XM_021848244.1 | XP_021703936.1 | 2 - 3 | 749 | 75 | ||
XM_021848245.1 | XP_021703937.1 | 2 - 3 | 626 | 75 | ||
XM_021848246.1 | XP_021703938.1 | 2 - 3 | 658 | 75 | ||
XM_021848247.1 | XP_021703939.1 | 2 - 3 | 704 | 75 | ||
XM_021848248.1 | XP_021703940.1 | 2 - 3 | 704 | 75 |
