Assay Details
Gene Symbol: LOC5564524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848242.1 | XP_021703934.1 | 8 - 9 | 4645 | 72 | |
XM_021848243.1 | XP_021703935.1 | 8 - 9 | 4780 | 72 | ||
XM_021848244.1 | XP_021703936.1 | 8 - 9 | 4690 | 72 | ||
XM_021848245.1 | XP_021703937.1 | 8 - 9 | 4567 | 72 | ||
XM_021848246.1 | XP_021703938.1 | 8 - 9 | 4599 | 72 | ||
XM_021848247.1 | XP_021703939.1 | 8 - 9 | 4630 | 72 | ||
XM_021848248.1 | XP_021703940.1 | 8 - 9 | 4645 | 72 |
