Assay Details
Gene Symbol: LOC5567482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852554.1 | XP_021708246.1 | 12 - 13 | 3417 | 93 | |
XM_021852555.1 | XP_021708247.1 | 12 - 13 | 3229 | 93 | ||
XM_021852556.1 | XP_021708248.1 | 12 - 13 | 3182 | 93 | ||
XM_021852557.1 | XP_021708249.1 | 12 - 13 | 3243 | 93 | ||
XM_021852558.1 | XP_021708250.1 | 12 - 13 | 3027 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
