Assay Details
Gene Symbol: LOC5567482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852554.1 | XP_021708246.1 | 11 - 12 | 3149 | 67 | |
XM_021852555.1 | XP_021708247.1 | 11 - 12 | 2961 | 67 | ||
XM_021852556.1 | XP_021708248.1 | 11 - 12 | 2914 | 67 | ||
XM_021852557.1 | XP_021708249.1 | 11 - 12 | 2975 | 67 | ||
XM_021852558.1 | XP_021708250.1 | 11 - 12 | 2759 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
