Assay Details
Gene Symbol: LOC5567482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852554.1 | XP_021708246.1 | 10 - 11 | 2960 | 71 | |
XM_021852555.1 | XP_021708247.1 | 10 - 11 | 2772 | 71 | ||
XM_021852556.1 | XP_021708248.1 | 10 - 11 | 2725 | 71 | ||
XM_021852557.1 | XP_021708249.1 | 10 - 11 | 2786 | 71 | ||
XM_021852558.1 | XP_021708250.1 | 10 - 11 | 2570 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
