Assay Details
Gene Symbol: LOC5567482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852554.1 | XP_021708246.1 | 3 - 4 | 1335 | 87 | |
XM_021852555.1 | XP_021708247.1 | 3 - 4 | 1147 | 87 | ||
XM_021852556.1 | XP_021708248.1 | 3 - 4 | 1100 | 87 | ||
XM_021852557.1 | XP_021708249.1 | 3 - 4 | 1161 | 87 | ||
XM_021852558.1 | XP_021708250.1 | 3 - 4 | 945 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
