Assay Details
Gene Symbol: LOC5578157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844714.1 | XP_021700406.1 | 4 - 5 | 1708 | 81 | |
XM_021844717.1 | XP_021700409.1 | 4 - 5 | 1302 | 81 |
