Assay Details
Gene Symbol: LOC5578157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844714.1 | XP_021700406.1 | 1 - 2 | 500 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021844714.1 transcripts.
Set Membership: |
![]() ![]() |
