Assay Details
Gene Symbol: LOC5564773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649078.2 | XP_001649128.2 | 3 - 4 | 1043 | 114 | |
XM_021851925.1 | XP_021707617.1 | 3 - 4 | 411 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
