Assay Details
Gene Symbol: LOC5564773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649078.2 | XP_001649128.2 | 5 - 6 | 1521 | 90 | |
XM_021851925.1 | XP_021707617.1 | 5 - 6 | 889 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
