Assay Details
Gene Symbol: LOC5567649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651793.2 | XP_001651843.2 | 1 - 2 | 267 | 84 | |
XM_021844642.1 | XP_021700334.1 | 1 - 2 | 267 | 84 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5567649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651793.2 | XP_001651843.2 | 1 - 2 | 267 | 84 | |
XM_021844642.1 | XP_021700334.1 | 1 - 2 | 267 | 84 |