Assay Details
Gene Symbol: LOC5567649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651793.2 | XP_001651843.2 | 3 - 4 | 891 | 94 | |
XM_021844638.1 | XP_021700330.1 | 4 - 5 | 1337 | 94 | ||
XM_021844639.1 | XP_021700331.1 | 3 - 4 | 1306 | 94 | ||
XM_021844641.1 | XP_021700333.1 | 4 - 5 | 1336 | 94 | ||
XM_021844642.1 | XP_021700334.1 | 3 - 4 | 891 | 94 |
