Assay Details
Gene Symbol: LOC5567649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651793.2 | XP_001651843.2 | 7 - 8 | 1532 | 82 | |
XM_021844641.1 | XP_021700333.1 | 8 - 9 | 1977 | 82 |
