Assay Details
Gene Symbol: LOC5567649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651793.2 | XP_001651843.2 | 8 - 9 | 1877 | 104 | |
XM_021844638.1 | XP_021700330.1 | 10 - 11 | 2329 | 104 | ||
XM_021844639.1 | XP_021700331.1 | 9 - 10 | 2298 | 104 | ||
XM_021844641.1 | XP_021700333.1 | 9 - 10 | 2322 | 104 | ||
XM_021844642.1 | XP_021700334.1 | 9 - 10 | 1883 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
