Assay Details
Gene Symbol: LOC5577010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857328.1 | XP_021713020.1 | 7 - 8 | 1594 | 64 | |
XM_021857329.1 | XP_021713021.1 | 7 - 8 | 1539 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
