Assay Details
Gene Symbol: LOC5577010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857328.1 | XP_021713020.1 | 6 - 7 | 1390 | 73 | |
XM_021857329.1 | XP_021713021.1 | 6 - 7 | 1335 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
