Assay Details
Gene Symbol: LOC5577010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857328.1 | XP_021713020.1 | 2 - 3 | 517 | 90 | |
XM_021857329.1 | XP_021713021.1 | 2 - 3 | 462 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021857328.1, XM_021857329.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
