Assay Details
Gene Symbol: LOC5570905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653333.2 | XP_001653383.2 | 10 - 11 | 2275 | 87 | |
XM_021844949.1 | XP_021700641.1 | 11 - 12 | 2352 | 87 | ||
XM_021844951.1 | XP_021700643.1 | 10 - 11 | 2204 | 87 | ||
XM_021844952.1 | XP_021700644.1 | 11 - 12 | 2372 | 87 | ||
XM_021844953.1 | XP_021700645.1 | 11 - 12 | 2371 | 87 | ||
XM_021844954.1 | XP_021700646.1 | 10 - 11 | 2186 | 87 | ||
XM_021844955.1 | XP_021700647.1 | 10 - 11 | 1998 | 87 | ||
XM_021844956.1 | XP_021700648.1 | 10 - 11 | 1784 | 87 | ||
XM_021844957.1 | XP_021700649.1 | 10 - 11 | 1719 | 87 | ||
XM_021844958.1 | XP_021700650.1 | 10 - 11 | 1792 | 87 | ||
XM_021844959.1 | XP_021700651.1 | 10 - 11 | 1732 | 87 |
