Assay Details
Gene Symbol: LOC5570905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653333.2 | XP_001653383.2 | 12 - 13 | 3021 | 68 | |
XM_021844949.1 | XP_021700641.1 | 13 - 14 | 3098 | 68 | ||
XM_021844951.1 | XP_021700643.1 | 12 - 13 | 2950 | 68 | ||
XM_021844952.1 | XP_021700644.1 | 13 - 14 | 3118 | 68 | ||
XM_021844953.1 | XP_021700645.1 | 13 - 14 | 3117 | 68 | ||
XM_021844954.1 | XP_021700646.1 | 12 - 13 | 2932 | 68 | ||
XM_021844955.1 | XP_021700647.1 | 12 - 13 | 2744 | 68 | ||
XM_021844956.1 | XP_021700648.1 | 12 - 13 | 2530 | 68 | ||
XM_021844957.1 | XP_021700649.1 | 12 - 13 | 2465 | 68 | ||
XM_021844958.1 | XP_021700650.1 | 12 - 13 | 2538 | 68 | ||
XM_021844959.1 | XP_021700651.1 | 12 - 13 | 2478 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
