Assay Details
Gene Symbol: LOC5571582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659741.2 | XP_001659791.1 | 1 - 2 | 553 | 86 | |
XM_021851752.1 | XP_021707444.1 | 1 - 2 | 553 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001659741.2, XM_021851752.1 transcripts.
Set Membership: |
![]() ![]() |
