Assay Details
Gene Symbol: LOC5569123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837656.1 | XP_021693348.1 | 4 - 5 | 1120 | 102 | |
XM_021837657.1 | XP_021693349.1 | 3 - 4 | 1021 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
