Assay Details
Gene Symbol: LOC5566218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650571.2 | XP_001650621.2 | 3 - 4 | 1406 | 73 | |
XM_021838781.1 | XP_021694473.1 | 3 - 4 | 976 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
