Assay Details
Gene Symbol: LOC5563827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848935.1 | XP_021704627.1 | 6 - 7 | 1724 | 63 | |
XM_021848937.1 | XP_021704629.1 | 6 - 7 | 1731 | 63 |

More Information
Set Membership: |
![]() ![]() |
